EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
An approach to 8 stereoisomers of homonojirimycin from (D)-glucose via kinetic & thermodynamic azido-γ-lactones.
PMID 23963282
Abstract
Crystal structures were obtained for the two C2 epimeric azido-γ-lactones 2-azido-2-deoxy-3,5:6,7-di-O-isopropylidene-d-glycero-d-ido-heptono-1,4-lactone and 2-azido-2-deoxy-3,5:6,7-di-O-isopropylidene-d-glycero-d-gulo-heptono-1,4-lactone prepared from kinetic and thermodynamic azide displacements of a triflate derived from d-glucoheptonolactone. Azido-γ-lactones are very useful intermediates in the synthesis of iminosugars and polyhydroxylated amino acids. In this study two epimeric azido-heptitols allow biotechnological transformations via Izumoring techniques to 8 of the 16 possible homonojirimycin analogues, 5 of which were isolated pure because of the lack of stereoselectivity of the final reductive amination. A side-by-side glycosidase inhibition profile of 11 of the possible 16 HNJ stereoisomers derived from d-glucose and d-mannose is presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
08012
1-Deoxynojirimycin, analytical standard
C6H13NO4