PloS one
MicroRNA-221 mediates the effects of PDGF-BB on migration, proliferation, and the epithelial-mesenchymal transition in pancreatic cancer cells.
PMID 23967190
Abstract
The platelet-derived growth factor (PDGF) signaling pathway has been found to play important roles in the development and progression of human cancers by regulating the processes of cell proliferation, apoptosis, migration, invasion, metastasis, and the acquisition of the epithelial-mesenchymal transition (EMT) phenotype. Moreover, PDGF signaling has also been found to alter the expression profile of miRNAs, leading to the reversal of EMT phenotype. Although the role of miRNAs in cancer has been documented, there are very few studies documenting the cellular consequences of targeted re-expression of specific miRNAs. Therefore, we investigated whether the treatment of human pancreatic cancer cells with PDGF could alter the expression profile of miRNAs, and we also assessed the cellular consequences. Our study demonstrates that miR-221 is essential for the PDGF-mediated EMT phenotype, migration, and growth of pancreatic cancer cells. Down-regulation of TRPS1 by miR-221 is critical for PDGF-mediated acquisition of the EMT phenotype. Additionally, the PDGF-dependent increase in cell proliferation appears to be mediated by inhibition of a specific target of miR-221 and down-regulation of p27Kip1.