EMAIL THIS PAGE TO A FRIEND
Langmuir : the ACS journal of surfaces and colloids
Palladium-catalyzed coupling reactions for the functionalization of Si surfaces: superior stability of alkenyl monolayers.
PMID 23968278
Abstract
Palladium-catalyzed Suzuki, Heck, and Sonogashira coupling reactions were studied as reaction protocols for organic modification of Si surfaces. These synthetically useful protocols allow for surface modification of alkene, alkyne, and halide terminated surfaces. Surface oxidation and metal contamination were assessed by X-ray photoelectron spectroscopy. The nature of the primary passivation layer was an important factor in the oxidation resistance of the Si surface during the secondary functionalization. Specifically, the use of alkynes as the primary functionalization layer gave superior stability compared to alkene analogues. The ability to utilize Pd-catalyzed coupling chemistries on Si surfaces opens great versatility for potential molecular and nanoscale electronics and sensing/biosensing applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
441201
3,4-Difluoroiodobenzene, 98%
C6H3F2I