EMAIL THIS PAGE TO A FRIEND
Anesthesiology
Medial septal cholinergic neurons modulate isoflurane anesthesia.
PMID 23969562
Abstract
Cholinergic drugs are known to modulate the response of general anesthesia. However, the sensitivity of isoflurane or other volatile anesthetics after selective lesion of septal cholinergic neurons that project to the hippocampus is not known. Male Long Evans rats had 192 immunoglobulin G-saporin infused into the medial septum (n = 10), in order to selectively lesion cholinergic neurons, whereas control, sham-lesioned rats were infused with saline (n = 12). Two weeks after septal infusion, the hypnotic properties of isoflurane and ketamine were measured using a behavioral endpoint of loss of righting reflex (LORR). Septal lesion was assessed by counting choline acetyltransferase-immunoreactive cells and parvalbumin-immunoreactive cells. Rats with 192 immunoglobulin G-saporin lesion, as compared with control rats with sham lesion, showed a 85% decrease in choline acetyltransferase-immunoreactive, but not parvalbumin-immunoreactive, neurons in the medial septal area. Lesioned as compared with control rats showed increased isoflurane sensitivity, characterized by a leftward shift of the graph plotting cumulative LORR percent with isoflurane dose. However, lesioned and control rats were not different in their LORR sensitivity to ketamine. When administered with 1.375% isoflurane, LORR induction time was shorter, whereas emergence time was longer, in lesioned as compared with control rats. Hippocampal 62-100 Hz gamma power in the electroencephalogram decreased with isoflurane dose, with a decrease that was greater in lesioned (n = 5) than control rats (n = 5). These findings suggest a role of the septal cholinergic neurons in modulating the sensitivity to isoflurane anesthesia, which affects both induction and emergence. The sensitivity of hippocampal gamma power to isoflurane appears to indicate anesthesia (LORR) sensitivity.