EMAIL THIS PAGE TO A FRIEND
Journal of pest science
Field resistance of
PMID 23970847
Abstract
The present studies were carried out to evaluate resistance in the populations of
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33969
Indoxacarb, PESTANAL®, analytical standard
C22H17ClF3N3O7
36159
Methomyl, PESTANAL®, analytical standard
C5H10N2O2S