EMAIL THIS PAGE TO A FRIEND
Bioelectrochemistry (Amsterdam, Netherlands)
Oxygen biosensor based on bilirubin oxidase immobilized on a nanostructured gold electrode.
PMID 23973738
Abstract
Gold disk electrodes modified with gold nanoparticles have been used as a scaffold for the covalent immobilization of bilirubin oxidase. The nanostructured bioelectrodes were tested as mediator-less biosensors for oxygen in a buffer that mimics the content and the composition of human physiological fluids. Chronoamperometry measurements showed a detection limit towards oxygen of 6 ± 1 μM with a linear range of 6-300 μM, i.e. exceeding usual physiological ranges of oxygen in human tissues and fluids. The biosensor presented is the first ever-reported oxygen amperometric biosensor based on direct electron transfer of bilirubin oxidase.