EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Magnetotactic bacteria form magnetite from a phosphate-rich ferric hydroxide via nanometric ferric (oxyhydr)oxide intermediates.
PMID 23980143
Abstract
The iron oxide mineral magnetite (Fe3O4) is produced by various organisms to exploit magnetic and mechanical properties. Magnetotactic bacteria have become one of the best model organisms for studying magnetite biomineralization, as their genomes are sequenced and tools are available for their genetic manipulation. However, the chemical route by which magnetite is formed intracellularly within the so-called magnetosomes has remained a matter of debate. Here we used X-ray absorption spectroscopy at cryogenic temperatures and transmission electron microscopic imaging techniques to chemically characterize and spatially resolve the mechanism of biomineralization in those microorganisms. We show that magnetite forms through phase transformation from a highly disordered phosphate-rich ferric hydroxide phase, consistent with prokaryotic ferritins, via transient nanometric ferric (oxyhydr)oxide intermediates within the magnetosome organelle. This pathway remarkably resembles recent results on synthetic magnetite formation and bears a high similarity to suggested mineralization mechanisms in higher organisms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
700312
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in toluene
Fe3O4
700320
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in toluene
Fe3O4
700304
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in toluene
Fe3O4
725331
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in H2O
Fe3O4
725358
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in H2O
Fe3O4
725366
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in H2O
Fe3O4
518158
Iron(II,III) oxide, 99.99% trace metals basis
Fe3O4
637106
Iron(II,III) oxide, nanopowder, 50-100 nm particle size (SEM), 97% trace metals basis
Fe3O4
310069
Iron(II,III) oxide, powder, <5 μm, 95%
Fe3O4
529311
Iron(III) oxide, ≥99.995% trace metals basis
Fe2O3
310050
Iron(III) oxide, powder, <5 μm, ≥96%
Fe2O3
544884
Iron(III) oxide, nanopowder, <50 nm particle size (BET)
Fe2O3
12344
Iron(III) oxide, puriss., ≥97.0% (complexometric)
Fe2O3
12342
Iron(III) oxide, purified, ≥95%
Fe2O3
720712
Iron(III) oxide, dispersion, nanoparticles, ≤110 nm particle size, 15 wt. % in ethanol
Fe2O3
720704
Iron(III) oxide, dispersion, nanoparticles, ≤110 nm particle size, 20 wt. % in H2O
Fe2O3