EMAIL THIS PAGE TO A FRIEND
Natural product communications
Development of a second generation palladium-catalyzed cycloalkenylation and its application to bioactive natural product synthesis.
PMID 23980435
Abstract
A novel palladium-catalyzed intramolecular oxidative alkylation of unactivated olefins is described. This protocol was devised to solve one of the drawbacks of the original palladium-catalyzed cycloalkenylation that we developed. We call this new procedure the 'second generation palladium-catalyzed cycloalkenylation'. This protocol has been applied to the total syntheses of cis-195A, trans-195A, boonein, scholareins A, C, D, and alpha-skytanthine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
203998
Palladium, granular, 99.99% trace metals basis
Pd
643181
Palladium, extent of labeling: 5 wt. % loading (dry basis), matrix activated carbon, wet support, Degussa type E105CA/W
Pd
326666
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Pd
203939
Palladium, powder, 99.995% trace metals basis
Pd
686468
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
Pd
267120
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Pd
348694
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Pd
267112
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Pd
464651
Palladium, powder, <75 μm, 99.9% trace metals basis
Pd
373206
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Pd
287474
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Pd
411450
Palladium, foil, thickness 0.05 mm, 99.9% trace metals basis
Pd
348643
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Pd
GF25547351
Palladium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.95%
Pd
GF68272588
Palladium, rod, 25mm, diameter 6.0mm, 99.95%
Pd
GF71618193
Palladium, wire reel, 5m, diameter 0.025mm, as drawn, 99.9%
Pd
GF05176122
Palladium, wire reel, 5m, diameter 0.05mm, as drawn, 99.9%
Pd
GF95863821
Palladium, foil, 50x50mm, thickness 0.1mm, as rolled, 99.95%
Pd
GF85752445
Palladium, foil, light tested, 25x25mm, thickness 0.006mm, 99.95%
Pd
GF03781863
Palladium, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 99.95%
Pd
GF50988358
Palladium, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 99.99+%
Pd
GF77561343
Palladium, foil, light tested, 25x25mm, thickness 0.03mm, as rolled, 99.95%
Pd
GF86358077
Palladium, foil, light tested, 25x25mm, thickness 0.047mm, as rolled, 99.95%
Pd
GF62160409
Palladium, foil, light tested, 25x25mm, thickness 0.075mm, as rolled, 99.95%
Pd
GF80658085
Palladium, foil, light tested, 50x50mm, thickness 0.025mm, as rolled, 99.95%
Pd
GF42154051
Palladium, foil, light tested, 50x50mm, thickness 0.025mm, as rolled, 99.99+%
Pd
GF36936399
Palladium, foil, light tested, 50x50mm, thickness 0.03mm, as rolled, 99.95%
Pd
GF94758991
Palladium, foil, light tested, 50x50mm, thickness 0.047mm, as rolled, 99.95%
Pd
GF89838220
Palladium, foil, light tested, 50x50mm, thickness 0.075mm, as rolled, 99.95%
Pd
GF51599425
Palladium, foil, not light tested, 100x100mm, thickness 0.004mm, 99.95%
Pd