EMAIL THIS PAGE TO A FRIEND
Lakartidningen
[New treatment options for vascular Ehlers-Danlos syndrome].
PMID 23980448
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000239 Celiprolol for peak identification, European Pharmacopoeia (EP) Reference Standard
C20H33N3O4 · HCl