EMAIL THIS PAGE TO A FRIEND
Seikagaku. The Journal of Japanese Biochemical Society
[Molecular mechanism of magnesium transport in renal tubule].
PMID 23980474
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
254118
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Mg
474754
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Mg
465992
Magnesium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Mg
254126
Magnesium, 20-230 mesh, reagent grade, 98%
Mg
63040
Magnesium, grit, ≥99.0% (KT)
Mg
253987
Magnesium, ReagentPlus®, powder, −50 mesh, ≥99%
Mg
465666
Magnesium, ReagentPlus®, powder, −325 mesh, 99.5% trace metals basis
Mg
13112
Magnesium, powder, ≥99%
Mg
266302
Magnesium, ReagentPlus®, ribbon, ≥99% trace metals basis
Mg
13103
Magnesium, ribbon, ≥99.5% Mg basis
Mg
299405
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Mg
403148
Magnesium, turnings, ~1/8 in., 99.95% trace metals basis
Mg
200905
Magnesium, turnings, reagent grade, 98%
Mg
63035
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Mg
723312
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Mg
723320
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Mg
735779
Magnesium, ingot, ≥99% trace metals basis
Mg
734365
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Mg
735760
Magnesium, plate, 99.95%
Mg
GF86050199
Magnesium, microfoil, 25x25mm, thinness 1.0μm, specific density 174μg/cm2, 6 micron aluminum permanent support, 99.9%
Mg
GF67975172
Magnesium, wire reel, 2m, diameter 1.1mm, as drawn, 99.9+%
Mg
GF80461964
Magnesium, microleaf, 25x25mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Mg
GF57746166
Magnesium, microleaf, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Mg
GF18270637
Magnesium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9+%
Mg
GF96495609
Magnesium, wire reel, 0.1m, diameter 0.25mm, as drawn, 99.9+%
Mg
GF04987547
Magnesium, wire reel, 0.1m, diameter 0.4mm, as drawn, 99.9+%
Mg
GF23637371
Magnesium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9+%
Mg
GF83427745
Magnesium, wire reel, 0.1m, diameter 1.0mm, as drawn, 99.9+%
Mg
GF86178189
Magnesium, wire reel, 0.1m, diameter 1.1mm, as drawn, 99.9+%
Mg
GF83829178
Magnesium, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.9+%
Mg