EMAIL THIS PAGE TO A FRIEND
Biochemistry
Thermodynamics and kinetics of adaptive binding in the malachite green RNA aptamer.
PMID 23984874
Abstract
Adaptive binding, the ability of molecules to fold themselves around the structure of a ligand and thereby incorporating it into their three-dimensional fold, is a key feature of most RNA aptamers. The malachite green aptamer (MGA) has been shown to bind several closely related triphenyl dyes with planar and nonplanar structures in this manner. Competitive binding studies using isothermal titration calorimetry and stopped flow kinetics have been conducted with the aim of understanding the adaptive nature of RNA-ligand interaction. The results of these studies reveal that binding of one ligand can reduce the ability of the aptamer pocket to adapt to another ligand, even if this second ligand has a significantly higher affinity to the free aptamer. A similar effect is observed in the presence of Mg(2+) ions which stabilize the binding pocket in a more ligand bound-like conformation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6158
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
C25H30ClN3
C0775
Crystal Violet, certified by the Biological Stain Commission
C25H30ClN3
46364
Crystal Violet, VETRANAL, analytical standard
C25H30ClN3
32675
Crystal Violet, indicator for the determination of the redox potential, S. No.: 785
C25H30ClN3
61135
Crystal Violet, for microscopy (Bact., Bot., Hist., Vit.), indicator (pH 0.1-2.0)
C25H30ClN3
HT90132
Crystal violet solution
C25H30ClN3
HT901
Crystal violet solution
C25H30ClN3
V5265
Crystal violet solution, 1%, aqueous solution
C25H30ClN3
G2039
Gentian Violet, meets USP testing specifications
C25H30ClN3
49144 Gentian Violet solution, for microscopy
M6880
Malachite Green oxalate salt, Technical grade
C23H25N2 · C2HO4 · 0.5C2H2O4
46396
Malachite Green oxalate salt, VETRANAL, analytical standard
C23H25N2 · C2HO4 · 0.5C2H2O4
M9015
Malachite Green oxalate salt, certified by the Biological Stain Commission
C23H25N2 · C2HO4 · 0.5C2H2O4
32745
Malachite Green oxalate salt, for microscopy, crystalline, S. No.: 754
C23H25N2 · C2HO4 · 0.5C2H2O4
38978
Malachite Green solution, for microscopy
P9172
Pyronin Y, for NA electrophoresis
C17H19ClN2O
83200
Pyronin Y, for microscopy (Bot., Fl., Hist.)
C17H19ClN2O
213519
Pyronin Y, certified by the Biological Stain Commission, Dye content 50 %
C17H19ClN2O
90903 Schaeffer and Fulton Spore Stain Solution A, for microscopy