EMAIL THIS PAGE TO A FRIEND
The American journal of clinical nutrition
Kynurenine pathway metabolites: relevant to vitamin B-6 deficiency and beyond.
PMID 23985806
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
148776
3-Hydroxyanthranilic acid, 97%
C7H7NO3