EMAIL THIS PAGE TO A FRIEND
Klinichna khirurhiia
[Caudal-epidural injection of adjuvants and local anaesthetics in the newborns undergoing surgery].
PMID 23987036
Abstract
In 45 newborns, operated on for the inborn failures of gut, the markers of stress and indices of central hemodynamics were studied up. General anaesthesia was applied together with caudal-epidural injection of local anaesthetics in 15 newborns, in 15--the adjuvants--klophelline and promedol--were added to local anaesthetics, in 15--a standard atharalgesia was used. The best and most durable effect was noted while application of anesthesia in combination with adjuvants.