EMAIL THIS PAGE TO A FRIEND
Journal of industrial microbiology & biotechnology
Efficient secretion of lipase r27RCL in Pichia pastoris by enhancing the disulfide bond formation pathway in the endoplasmic reticulum.
PMID 23990169
Abstract
The lipase r27RCL from Rhizopus chinensis CCTCC M201021 was heterologously expressed in Pichia pastoris GS115 by simultaneous co-expression with two secretion factors ERO1p and PDI involved in the endoplasmic reticulum (ER). Compared to the expression of the lipase alone (12,500 U/ml), co-expression with these two proteins resulted in the production of larger total quantities of enzymes. The largest increase was seen when the combined ERO1p/PDI system was co-expressed, resulting in approximately 30 % higher enzyme yields (16,200 U/ml) than in the absence of co-expressed secretion factors. The extracellular protein concentration of the recombinant strain Co XY RCL-5 reached 9.39 g/l in the 7-l fermentor. Simultaneously, the fermentation time was also shortened by about 8 h compared to that of the control. The substrate-specific consumption rate (Qs) and the product-specific production rate (Qp) were both investigated in this research. In conclusion, the space-time yield was improved by co-expression with ERO1p and PDI. This is a potential strategy for high level expression of other heterologous proteins in P. pastoris.