EMAIL THIS PAGE TO A FRIEND
Journal of lipid research
Cooperation between hepatic cholesteryl ester hydrolase and scavenger receptor BI for hydrolysis of HDL-CE.
PMID 23990661
Abstract
Liver is the sole organ responsible for the final elimination of cholesterol from the body either as biliary cholesterol or bile acids. High density lipoprotein (HDL)-derived cholesterol is the major source of biliary sterols and represents a mechanism for the removal of cholesterol from peripheral tissues including artery wall-associated macrophage foam cells. Via selective uptake through scavenger receptor BI (SR-BI), HDL-cholesterol is thought to be directly secreted into bile, and HDL cholesteryl esters (HDL-CEs) enter the hepatic metabolic pool and need to be hydrolyzed prior to conversion to bile acids. However, the identity of hepatic CE hydrolase (CEH) as well as the role of SR-BI in bile acid synthesis remains elusive. In this study we examined the role of human hepatic CEH (CES1) in facilitating hydrolysis of SR-BI-delivered HDL-CEs. Over-expression of CEH led to increased hydrolysis of HDL-[³H]CE in primary hepatocytes and SR-BI expression was required for this process. Intracellular CEH associated with BODIPY-CE delivered by selective uptake via SR-BI. CEH and SR-BI expression enhanced the movement of [³H]label from HDL-[³H]CE to bile acids in vitro and in vivo. Taken together, these studies demonstrate that SR-BI-delivered HDL-CEs are hydrolyzed by hepatic CEH and utilized for bile acid synthesis.