EMAIL THIS PAGE TO A FRIEND
eLife
Patient-specific iPSC-derived photoreceptor precursor cells as a means to investigate retinitis pigmentosa.
PMID 23991284
Abstract
Next-generation and Sanger sequencing were combined to identify disease-causing USH2A mutations in an adult patient with autosomal recessive RP. Induced pluripotent stem cells (iPSCs), generated from the patient's keratinocytes, were differentiated into multi-layer eyecup-like structures with features of human retinal precursor cells. The inner layer of the eyecups contained photoreceptor precursor cells that expressed photoreceptor markers and exhibited axonemes and basal bodies characteristic of outer segments. Analysis of the USH2A transcripts of these cells revealed that one of the patient's mutations causes exonification of intron 40, a translation frameshift and a premature stop codon. Western blotting revealed upregulation of GRP78 and GRP94, suggesting that the patient's other USH2A variant (Arg4192His) causes disease through protein misfolding and ER stress. Transplantation into 4-day-old immunodeficient Crb1 (-/-) mice resulted in the formation of morphologically and immunohistochemically recognizable photoreceptor cells, suggesting that the mutations in this patient act via post-developmental photoreceptor degeneration. DOI:http://dx.doi.org/10.7554/eLife.00824.001.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB360
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5 detects level of Glial Fibrillary Acidic Protein & has been published & validated for use in IC, IH, IH(P) & WB with more than 65 product citations.
AB5407
Anti-Opsin Antibody, blue, This Anti-Opsin Antibody, blue is validated for use in IH(P) for the detection of Opsin.
AB5405
Anti-Opsin Antibody, Red/Green, Anti-Opsin Antibody, Red/Green detects level of Opsin & has been published & validated for use in IH(P).
MAB5552
Anti-Pax6 Antibody, Detect Pax6 using this Anti-Pax6 Antibody validated for use in IH(P) & WB.
AB5585
Anti-Recoverin Antibody, Detect Recoverin using this Anti-Recoverin Antibody validated for use in IC, IH, IH(P) & WB.
MAB5428
Anti-Retinal Pigment Epithelium 65 Antibody, Anti-Retinal Pigment Epithelium 65 Antibody detects level of Retinal Pigment Epithelium 65 & has been published & validated for use in ELISA, IH, IP & WB.
MAB5316
Anti-Rhodopsin Antibody, clone RET-P1, Detect Rhodopsin using this Anti-Rhodopsin Antibody, clone RET-P1 validated for use in IC & WB.
MAB4385
Anti-TRA-1-85 Antibody, blood group Antigen Ok(a), clone TRA-1-85, Anti-TRA-1-85 Antibody, blood group antigen Ok(a), clone TRA-1-85 detects level of TRA-1-85 & has been published & validated for use in FC, IP & WB.
MABT11
Anti-ZO-1 Antibody, clone R40.76, Anti-ZO-1 Antibody, clone R40.76 detects level of ZO-1 & has been published & validated for use in WB, IP, IH.
G8022
Gastrin Releasing Peptide human, ≥97% (HPLC)
C130H204N38O31S2