EMAIL THIS PAGE TO A FRIEND
Journal of the New Jersey Dental Association
Platelets and bleeding in the dental patient. It's not always from "blood thinners". Von Willebrand disease--clinical assessment and case report.
PMID 23991505