EMAIL THIS PAGE TO A FRIEND
Nanotoxicology
Vasoactive effects of stable aqueous suspensions of single walled carbon nanotubes in hamsters and mice.
PMID 23992463
Abstract
Single-walled carbon nanotubes synthesized with iron (Fe-SWCNT) or gadolinium (Gd-SWCNT) show promise as high performance multimodal contrast and drug-delivery agents. Our purpose was to evaluate potential vasoactive effects of SWCNT. Stable aqueous solutions of Fe-SWCNTs or Gd-SWCNTs were made using the biocompatible amphiphilic polymer N-(carbonyl-methoxypolyethyleneglycol 2000)-1,2-distearoylsn-glycero-3- phosphoethanolamine (PEG-DSPE). Both aggregated and non-aggregated (sonicated) formulations were tested. The initial vasoactivity of the formulations and their potential for inducing pro-inflammatory endothelial dysfunction were investigated in the hamster cheek pouch and murine cremaster muscle intravital microscopy models. These models provide an assay to test several formulations/dosages in a paired fashion. Abluminal exposure to small arterioles exposes both endothelial and vascular smooth muscle cells. Using abluminal exposures of dosages that would approximate the first pass of an i.v. bolus injection, both Fe-SWCNTs and Gd-SWCNTs were immediately vasoactive. Aggregated formulations induced dilation and non-aggregated formulations induced constriction in both hamsters and mice. Endothelial dysfunction was evident after exposure to either aggregated or non-aggregated forms. General loss of dilator capability was seen after exposure to non-aggregated but not aggregated forms. Thus concentrations mimicking bolus dosing of PEG-DSPE coated SWCNT induce both acute and chronic vascular responses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF58945078
Iron, wire reel, 250m, diameter 0.25mm, hard, 99.5%
Fe
GF21695722
Iron, wire reel, 2m, diameter 0.13mm, hard, 99.99+%
Fe
GF81282135
Iron, wire reel, 2m, diameter 0.25mm, annealed, 99.99+%
Fe
GF57565996
Iron, wire reel, 2m, diameter 0.25mm, hard, 99.99+%
Fe
GF48022152
Iron, wire reel, 2m, diameter 0.5mm, hard, 99.99+%
Fe
GF48020947
Iron, wire reel, 500m, diameter 0.125mm, hard, 99.5%
Fe
GF58809965
Iron, wire reel, 50m, diameter 0.065mm, hard, 99.5%
Fe
GF34071566
Iron, wire reel, 50m, diameter 0.25mm, hard, 99.5%
Fe
GF96915134
Iron, wire reel, 50m, diameter 0.5mm, as drawn, 99.95%
Fe
GF35663935
Iron, wire reel, 5m, diameter 0.025mm, as drawn, 99.99+%
Fe
GF27511781
Iron, wire reel, 5m, diameter 0.25mm, annealed, 99.99+%
Fe
GF61040023
Iron, wire reel, 5m, diameter 0.25mm, hard, 99.99+%
Fe
GF55306964
Iron, wire reel, 5m, diameter 0.5mm, hard, 99.99+%
Fe
GF92087242
Iron, wire reel, 5m, diameter 0.5mm, as drawn, 99.95%
Fe
GF83094824
Iron, rod, 300mm, diameter 6.0mm, as drawn, 99.99+%
Fe
GF97671415
Iron, rod, 500mm, diameter 4.8mm, as drawn, 98+%
Fe
GF19311724
Iron, rod, 500mm, diameter 9.5mm, as drawn, 98+%
Fe
GF91822653
Iron, rod, 50mm, diameter 40mm, as drawn, armcO« soft ingot 99.8%
Fe
GF39505518
Iron, tube, 1000mm, outside diameter 1.40mm, inside diameter 0.8mm, wall thickness 0.3mm, as drawn, 99.8%
Fe
GF62180928
Iron, wire reel, 2000m, diameter 0.065mm, hard, 99.5%
Fe
GF05699975
Iron, wire reel, 250m, diameter 0.065mm, hard, 99.5%
Fe
GF56751206
Iron, wire reel, 2m, diameter 1.0mm, as drawn, 99.99+%
Fe
GF01815702
Iron, wire reel, 5m, diameter 0.13mm, hard, 99.99+%
Fe
GF17042082
Iron, tube, 200mm, outside diameter 4.0mm, inside diameter 3.6mm, wall thickness 0.2mm, as drawn, 99.5%
Fe
GF06733818
Iron, tube, 200mm, outside diameter 5.0mm, inside diameter 4.5mm, wall thickness 0.25mm, as drawn, 99.5%
Fe
GF45478604
Iron, tube, 200mm, outside diameter 6.35mm, inside diameter 5.35mm, wall thickness 0.5mm, as drawn, 99.8%
Fe
GF93209328
Iron, tube, 200mm, outside diameter 8.0mm, inside diameter 5mm, wall thickness 1.5mm, annealed, 99.5%
Fe
GF68348076
Iron, tube, 200mm, outside diameter 9.5mm, inside diameter 8.5mm, wall thickness 0.5mm, as drawn, 99.8+%
Fe
GF55566816
Iron, tube, 500mm, outside diameter 1.0mm, inside diameter 0.8mm, wall thickness 0.1mm, as drawn, 99.5%
Fe
GF78871988
Iron, tube, 500mm, outside diameter 1.40mm, inside diameter 0.8mm, wall thickness 0.3mm, as drawn, 99.8%
Fe