EMAIL THIS PAGE TO A FRIEND
The Journal of allergy and clinical immunology
Tissue transglutaminase generates deamidated epitopes on gluten, increasing reactivity with hydrolyzed wheat protein-sensitized IgE.
PMID 23992746