EMAIL THIS PAGE TO A FRIEND
Fertility and sterility
Successful testicular sperm retrieval in adolescents with Klinefelter syndrome treated with at least 1 year of topical testosterone and aromatase inhibitor.
PMID 23993925