EMAIL THIS PAGE TO A FRIEND
Translational research : the journal of laboratory and clinical medicine
The serum level of soluble CD26/dipeptidyl peptidase 4 increases in response to acute hyperglycemia after an oral glucose load in healthy subjects: association with high-molecular weight adiponectin and hepatic enzymes.
PMID 23994650
Abstract
A soluble form of CD26/dipeptidyl peptidase 4 (sCD26/DPP4) is found in serum and it has DPP4 enzymatic activity. We investigated whether the serum level of sCD26/DPP4 was influenced by the oral glucose tolerance test (OGTT) in healthy subjects. The serum sCD26/DPP4 level increased significantly from 824.5 ng/mL (interquartile range, from 699.0 to 1050 ng/mL) at baseline to a peak of 985.0 ng/mL (interquartile range, from 796.5 to 1215 ng/mL) during the OGTT (P < 0.0001). The peak sCD26/DPP4 level correlated positively with the baseline age and body mass index, and fasting plasma glucose (FPG), homeostasis model assessment of insulin resistance (HOMA-IR), triglycerides (TG), alanine aminotransferase, and γ-glutamyl transpeptidase (GGT) levels whereas it correlated negatively with high-density lipoprotein (HDL) cholesterol and the serum levels of total and high-molecular weight (HMW) adiponectin. Stepwise regression analysis was done with forward selection of variables, including age, FPG, HOMA-IR, TG, HDL cholesterol, uric acid, GGT, C-reactive protein, and HMW adiponectin. In a model that explained 57.5% of the variation of the peak sCD26/DPP4 level, GGT (β = 0.382, P = 0.007) and HOMA-IR (β = 0.307, P = 0.034) were independent determinants of the peak serum level of sCD26/DPP4. Serum HMW adiponectin decreased significantly from 4.43 μg/mL (interquartile range, from 2.80 to 6.65 μg/mL) at baseline to 4.17 μg/mL (interquartile range, from 2.48 to 6.96 μg/mL) 120 minutes after the oral glucose load (P < 0.0001). The baseline serum level of sCD26/DPP4 showed a significant negative correlation with the percent change of HMW adiponectin during the OGTT. In conclusion, the serum level of sCD26/DPP4 increased acutely after an oral glucose load in apparently healthy subjects. The abrupt increase of serum sCD26/DPP4 after a glucose load may be a marker of insulin resistance that could come from liver or muscle.