EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Efficient direct oxygen reduction by laccases attached and oriented on pyrene-functionalized polypyrrole/carbon nanotube electrodes.
PMID 23994955
Abstract
We report the functionalization of multi-walled carbon nanotube (MWCNT) electrodes by oxidative electropolymerization of pyrrole monomers bearing pyrene and N-hydroxysuccinimide groups. Both polymers were applied to the immobilization and electrical wiring of Trametes versicolor laccase via chemical grafting or non-covalent binding. A "pseudo" host-guest interaction of polymerized pyrene with a hydrophobic cavity of laccase was used for the oriented enzyme immobilization on MWCNT electrodes. The latter leads to higher catalytic current for oxygen reduction (1.85 mA cm(-2)) and higher electroenzymatic stability (50% after one month).