EMAIL THIS PAGE TO A FRIEND
Anesteziologiia i reanimatologiia
[The use of central acting analgesic nefopam in postoperative analgesia in cardiac surgery patients].
PMID 24000658
Abstract
A prospective, randomized, comparative study was conducted. 3 analgesia protocols were used: 1) patient controlled analgesia (PCA) with trimeperidine in combination with a nefopam constant infusion; 2) PCA with trimeperidine in combination with a nefopam bolus; 3) PCA with trimeperidine separately during early postoperative period in cardiac surgery patients. The study included 60 patients agedf rom 40 to 65 years of age (20 patients in each group). The analgesia efficacy was evaluated with a 5-point verbal rating scale (VRS) for pain intensity and inspiratory lung capacity (ILC), measured with incentive spirometer. The safety of nefopam during early postoperative period in cardiac surgery patients was shown. The combination of nefopam and trimeperidine led to a more pronounced analgetic effect. Trimeperidine consumption was significantly lower in nefopam groups than in the group of isolated PCA. Wholly adverse effects were associated with trimeperidine and were dose-related The incidence of nausea, vomiting, dizziness, weakness, bowel paresis was significantly higher in isolated PCA group than in the other two groups.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0909
Nefopam hydrochloride, ≥98% (HPLC)
C17H19NO · HCl