EMAIL THIS PAGE TO A FRIEND
Journal of AOAC International
Multivariate optimization of separation conditions for simultaneous determination of acemetacin and chlorzoxazone in a pharmaceutical preparation by HPLC using response surface methodology.
PMID 24000743
Abstract
A new, fast, accurate, precise, and sensitive RP-HPLC method for the simultaneous determination of acemetacin and chlorzoxazone has been developed. Response surface methodology with a central composite design was used to optimize the acetonitrile and ammonium acetate percentage in the mobile phase and pH of ammonium acetate. The optimum separation was achieved on a C18 column (250 x 4.6 mm id, 5 microm particle size) using the mobile phase methanol-acetonitrile-0.02 M ammonium acetate, pH 9.4 (25 + 35 + 40, v/v/v) at a flow rate of 1.5 mL/min; UV detection at 270 nm, and cyanocobalamin as an internal standard. This developed method was validated and successfully applied to a coated tablet pharmaceutical preparation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1674
Acemetacin, analytical standard
C21H18ClNO6