EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
On-line two-dimensional capillary strong anion exchange/reversed phase liquid chromatography-tandem mass spectrometry for comprehensive lipid analysis.
PMID 24001943