EMAIL THIS PAGE TO A FRIEND
Eksperimental'naia i klinicheskaia farmakologiia
[The effect of betahistine on histological changes in rabbit brain in model of whole body wide-frequency vibration].
PMID 24003483
Abstract
In acute experiments in conscious rabbits was studied protective action of selective blocker of histamine H3-receptor betahistine (2mg/kg i/v) against histological changes in precentral and postcentral gyrus, as well as in temporal lobe of cerebral cortex, thalamus, hypothalamus, and cerebellum, arising in case of modeling of whole body wide-frequency vibration. Betahistine attenuates edematous and degenerative changes in neurons and reciprocal glial reaction, caused by vibration, but does not eliminate edema in perivascular spaces. This effect may be related to the improvement of blood supply as a result of of vasodilatory action and decrease of oxygen consumption via vestibuloprotective effect.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B0990000 Betahistine mesilate, European Pharmacopoeia (EP) Reference Standard
C8H12N2 · 2CH4O3S