EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Cell physiology
Sickle cell anemia: time for personalized prescription of hydroxyurea? Focus on "Organic anion transporting polypeptide 1B transporters modulate hydroxyurea pharmacokinetics".
PMID 24005044