EMAIL THIS PAGE TO A FRIEND
Giornale italiano di dermatologia e venereologia : organo ufficiale, Societa italiana di dermatologia e sifilografia
A case of skin and lung tuberculosis: diagnostic difficulties.
PMID 24005149
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E4630
Ethambutol dihydrochloride, antimycobacterial
C10H24N2O2 · 2HCl