EMAIL THIS PAGE TO A FRIEND
BioTechniques
Improved chemiluminescent detection of alkaline phosphatase.
PMID 2400598
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31589
Diethanolamine, BioUltra, ≥99.5% (GC)
C4H11NO2