EMAIL THIS PAGE TO A FRIEND
Organic letters
γ-Substituted butanolides from cyclopropane hemimalonates: an expedient synthesis of natural (R)-dodecan-4-olide.
PMID 24007510
Abstract
Exploration into the reactivity of donor-acceptor cyclopropane hemimalonates has led to the facile synthesis of γ-substituted butanolides. Under microwave irradiation, cyclopropane hemimalonates undergo rapid conversion to butanolides in the presence of inorganic salts with an unprecedented retention of stereochemistry. This unique process has been applied to the total synthesis of the naturally occurring (R)-dodecan-4-olide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295183
Cyclopropane, ≥99%
C3H6
20740
γ-Butyrolactone, puriss., ≥99.0% (GC)
C4H6O2