EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Antitrypanosomal triterpenoid with an ε-lactone E-ring from Salvia urmiensis.
PMID 24007549
Abstract
A new triterpenoid, urmiensolide (1), was isolated from Salvia urmiensis. The structure was elucidated by a combination of 1D and 2D NMR, HRESIMS, and X-ray crystallographic analyses. The absolute configuration was established by comparison of experimental and simulated ECD spectra. Urmiensolide is the first pentacyclic triterpenoid bearing a ε-lactone E-ring. The compound showed in vitro antitrypanosoal activity with an IC₅₀ value of 5.6 μM against the Trypanosoma brucei rhodesiense STIB 900 strain and a selectivity index of 33. A possible biosynthetic pathway of 1 from α-amyrin is proposed.