EMAIL THIS PAGE TO A FRIEND
International journal of oncology
Hematein, a casein kinase II inhibitor, inhibits lung cancer tumor growth in a murine xenograft model.
PMID 24008396
Abstract
Casein kinase II (CK2) inhibitors suppress cancer cell growth. In this study, we examined the inhibitory effects of a novel CK2 inhibitor, hematein, on tumor growth in a murine xenograft model. We found that in lung cancer cells, hematein inhibited cancer cell growth, Akt/PKB Ser129 phosphorylation, the Wnt/TCF pathway and increased apoptosis. In a murine xenograft model of lung cancer, hematein inhibited tumor growth without significant toxicity to the mice tested. Molecular docking showed that hematein binds to CK2α in durable binding sites. Collectively, our results suggest that hematein is an allosteric inhibitor of protein kinase CK2 and has antitumor activity to lung cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
51230
Hematein, for microscopy (Hist.)
C16H12O6