EMAIL THIS PAGE TO A FRIEND
Journal of the American Society of Nephrology
Reduced ciliary polycystin-2 in induced pluripotent stem cells from polycystic kidney disease patients with PKD1 mutations.
PMID 24009235