EMAIL THIS PAGE TO A FRIEND
Ugeskrift for laeger
[Ketamine for treatment of acute depression].
PMID 24011203
Abstract
In clinical trials a single dose of the N-methyl-D-aspartate (NMDA) receptor antagonist ketamine has shown a rapid antidepressant effect in patients with treatment-resistant depression and bipolar depression. The implications of glutaminergic mechanisms in depression and the rapid effect of a single dose of ketamine could open new pathways to understand the pathophysiology of depression and the development of novel rapid-acting antidepressant drugs.