EMAIL THIS PAGE TO A FRIEND
American journal of kidney diseases : the official journal of the National Kidney Foundation
Vitamin A treatment for psoriasis vulgaris in a dialysis patient.
PMID 24011973
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1011029 Acitretin Related Compound B, United States Pharmacopeia (USP) Reference Standard
C23H30O3