EMAIL THIS PAGE TO A FRIEND
Journal of bioscience and bioengineering
Effect of growth rate on plasmid DNA production and metabolic performance of engineered Escherichia coli strains.
PMID 24012107
Abstract
Two engineered Escherichia coli strains, designated VH33 and VH34, were compared to their parent strain W3110 in chemostat mode during plasmid DNA (pDNA) production. In strain VH33 the glucose uptake system was modified with the aim of reducing overflow metabolism. The strain VH34 has an additional deletion of the pyruvate kinase A gene (pykA) to increase pDNA formation. pDNA formation rates as well as kinetic and stoichiometric parameters were investigated in dependence of the growth rate within a range from 0.02 to 0.25 h(-1). Differences between strains were found in terms of the biomass yields on nitrogen and oxygen, as well as on the cell maintenance coefficients. The deletion of pykA led to a significantly increased pDNA yield and productivity. At an optimal growth rate of 0.20 h(-1) it was nearly 60% higher than that of W3110 and VH33. Metabolic fluxes calculated by metabolite balance analysis showed differences mainly in reactions catalyzed by pyruvate kinase and glucose 6-phosphate dehydrogenase. The obtained data are useful for the design of cultivation schemes for pDNA production by E. coli.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G6378 Glucose-6-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), Type XV, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
G7877 Glucose-6-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), Type VII, ammonium sulfate suspension, ≥200 units/mg protein
G4134 Glucose-6-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), Type IX, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
G5885 Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, lyophilized powder, 550-1,100 units/mg protein (biuret)
G8404 Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, ammonium sulfate suspension, 550-1,100 units/mg protein (biuret)
G5760 Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, Type XXIII, ammonium sulfate suspension, 550-1,100 units/mg protein (biuret), ≥2.0 mg/mL Biuret
G8529 Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, lyophilized powder, 550-1,100 units/mg protein (biuret)
P1903 Pyruvate Kinase from Bacillus stearothermophilus, Type VIII, lyophilized powder, 100-300 units/mg protein
P1506 Pyruvate Kinase from rabbit muscle, Type II, ammonium sulfate suspension, 350-600 units/mg protein
P9136 Pyruvate Kinase from rabbit muscle, Type III, lyophilized powder, 350-600 units/mg protein
P7768 Pyruvate Kinase from rabbit muscle, Type VII, buffered aqueous glycerol solution, 350-600 units/mg protein