EMAIL THIS PAGE TO A FRIEND
Cell cycle (Georgetown, Tex.)
Nuclear PKM2 regulates the Warburg effect.
PMID 24013426
Abstract
Pyruvate kinase is a rate-limiting glycolytic enzyme. The PKM1 and PKM2 isoforms result from mutually exclusive alternative splicing of the PKM pre-mRNA. PKM2 rather than PKM1 regulates the Warburg effect and tumorigenesis by poorly understood mechanisms. Emerging evidence has revealed that ERK1/2 phosphorylates PKM2, but not PKM1, leading to PIN1-dependent cis-trans isomerization and conversion of PKM2 from a tetramer to a monomer. Monomeric PKM2 translocates into the nucleus, where it functions as a histone kinase and upregulates the expression of c-Myc and cyclin D1, thereby promoting the Warburg effect and cell cycle progression, respectively. Thus, nuclear PKM2 is essential for tumorigenesis and may serve as a target for treating human cancer.