EMAIL THIS PAGE TO A FRIEND
Eur. J. Drug Metab. Pharmacokinet.
Changes in the expression levels of tight junction components during reconstruction of tight junction from mucosal lesion by intestinal ischemia/reperfusion.
PMID 24014129