EMAIL THIS PAGE TO A FRIEND
ACS combinatorial science
One-pot, two-step synthesis of imidazo[1,2-a]benzimidazoles via a multicomponent [4 + 1] cycloaddition reaction.
PMID 24016144
Abstract
A one-pot, two-step synthesis of imidazo[1,2-a]benzimidazoles has been achieved by a three-component reaction of 2-aminobenzimidazoles with an aromatic aldehyde and an isocyanide. The reaction involving condensation of 2-aminobenzimidazole with an aldehyde is run under microwave activation to generate an imine intermediate under basic conditions which then undergoes [4 + 1] cycloaddition with an isocyanide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
171778
2-Aminobenzimidazole, 97%
C7H7N3