EMAIL THIS PAGE TO A FRIEND
Organic letters
Regioselectivity of the oxidative C-S bond formation in ergothioneine and ovothiol biosyntheses.
PMID 24016264
Abstract
Ergothioneine (5) and ovothiol (8) are two novel thiol-containing natural products. Their C-S bonds are formed by oxidative coupling reactions catalyzed by EgtB and OvoA enzymes, respectively. In this work, it was discovered that in addition to catalyzing the oxidative coupling between histidine and cysteine (1 → 6 conversion), OvoA can also catalyze a direct oxidative coupling between hercynine (2) and cysteine (2 → 4 conversion), which can shorten the ergothioneine biosynthetic pathway by two steps.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B3501
Betaine hydrochloride, ≥99%
C5H12ClNO2
E7521
L-(+)-Ergothioneine
C9H15N3O2S