EMAIL THIS PAGE TO A FRIEND
Organic letters
Nonaggregational shape-persistent cyclo[6]aramide and its macrocyclic effect toward binding secondary ammonium salts in moderately polar media.
PMID 24024875
Abstract
Simply by introducing steric side chains, the shape-persistent cyclo[6]aramides were found to exhibit nonaggregational behavior and strong association (3 × 10(4) M(-1)) ability in acetone for binding secondary ammonium salt. The complexation can be switched in an on-and-off fashion using AgPF6 and TBACl, contrasting sharply with their corresponding acyclic pentamer and demonstrating the macrocyclic effect.