EMAIL THIS PAGE TO A FRIEND
Drug Development and Industrial Pharmacy
Hydrogel-gauze dressing for moderate-to-severe atopic dermatitis: development and efficacy study on atopic dermatitis-like skin lesions in NC/Nga mice.
PMID 24025072