EMAIL THIS PAGE TO A FRIEND
The Journal of neuropsychiatry and clinical neurosciences
Donepezil-induced confusional state in a patient with autopsy-proven behavioral-variant frontotemporal dementia.
PMID 24026732
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1224981 Donepezil hydrochloride, United States Pharmacopeia (USP) Reference Standard
C24H29NO3 · HCl