EMAIL THIS PAGE TO A FRIEND
International journal of STD & AIDS
Acute generalised exanthematous pustulosis induced by Pneumocystis jirovecii pneumonia prophylaxis with dapsone.
PMID 24026795
Abstract
We describe the case of HIV-1 infected patient presenting to hospital with a severe cutaneous adverse drug reaction shortly after commencing dapsone therapy as Pneumocystis jirovecii pneumonia prophylaxis. To the best of our knowledge, acute generalised exanthematous pustulosis has not been reported as a reaction to dapsone in the setting of HIV.