EMAIL THIS PAGE TO A FRIEND
Acta neurologica Taiwanica
Intrasinus thrombolysis by mechanical and urokinase for severe cerebral venous sinus thrombosis : a case report.
PMID 24030091
Abstract
Cerebral venous and sinus thrombosis (CVST) is a rare stroke disorder, which requires prompt recognition and appropriate intervention to prevent a devastating outcome. Intrasinus thrombolysis is an alternative and effective method to anticoagulant therapy for the treatment of CVST, but is rarely used in Taiwan. A 46-year-old man presented with a one-week history of intractable headache and progressive weakness of his right lower limb. Magnetic resonance venography of the brain confirmed a diagnosis of extensive venous sinus thrombosis. The patient was successfully treated by direct intrasinus thrombolysis with urokinase. This case highlights the benefit of early intrasinus thrombolysis for the treatment of CVST.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
U4010 Urokinase from human kidney cells, lyophilized powder