EMAIL THIS PAGE TO A FRIEND
American journal of respiratory and critical care medicine
The effect of etanercept on lung leukocyte margination and fibrin deposition after cardiac surgery.
PMID 24032386
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5386 Fibrin from human plasma, insoluble powder