EMAIL THIS PAGE TO A FRIEND
American journal of human genetics
Mutations in GMPPA cause a glycosylation disorder characterized by intellectual disability and autonomic dysfunction.
PMID 24035193
Abstract
In guanosine diphosphate (GDP)-mannose pyrophosphorylase A (GMPPA), we identified a homozygous nonsense mutation that segregated with achalasia and alacrima, delayed developmental milestones, and gait abnormalities in a consanguineous Pakistani pedigree. Mutations in GMPPA were subsequently found in ten additional individuals from eight independent families affected by the combination of achalasia, alacrima, and neurological deficits. This autosomal-recessive disorder shows many similarities with triple A syndrome, which is characterized by achalasia, alacrima, and variable neurological deficits in combination with adrenal insufficiency. GMPPA is a largely uncharacterized homolog of GMPPB. GMPPB catalyzes the formation of GDP-mannose, which is an essential precursor of glycan moieties of glycoproteins and glycolipids and is associated with congenital and limb-girdle muscular dystrophies with hypoglycosylation of α-dystroglycan. Surprisingly, GDP-mannose pyrophosphorylase activity was unchanged and GDP-mannose levels were strongly increased in lymphoblasts of individuals with GMPPA mutations. This suggests that GMPPA might serve as a GMPPB regulatory subunit mediating feedback inhibition of GMPPB instead of displaying catalytic enzyme activity itself. Thus, a triple-A-like syndrome can be added to the growing list of congenital disorders of glycosylation, in which dysregulation rather than mere enzyme deficiency is the basal pathophysiological mechanism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV49209
Anti-GMPPA antibody produced in rabbit, affinity isolated antibody