EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
Nickel-dependent metalloenzymes.
PMID 24036122
Abstract
This review describes the functions, structures, and mechanisms of nine nickel-containing enzymes: glyoxalase I, acireductone dioxygenase, urease, superoxide dismutase, [NiFe]-hydrogenase, carbon monoxide dehydrogenase, acetyl-coenzyme A synthase/decarbonylase, methyl-coenzyme M reductase, and lactate racemase. These enzymes catalyze their various chemistries by using metallocenters of diverse structures, including mononuclear nickel, dinuclear nickel, nickel-iron heterodinuclear sites, more complex nickel-containing clusters, and nickel-tetrapyrroles. Selected other enzymes are active with nickel, but the physiological relevance of this metal specificity is unclear. Additional nickel-containing proteins of undefined function have been identified.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G4252 Glyoxalase I from Saccharomyces cerevisiae, Grade IV, buffered aqueous glycerol solution, ≥400 units/mg protein
GF82156912
Nickel, rod, 100mm, diameter 12.0mm, 99%
Ni
GF11598194
Nickel, rod, 100mm, diameter 12.7mm, 99.98%
Ni
GF29576010
Nickel, rod, 100mm, diameter 16mm, 99.99+%
Ni
GF83584143
Nickel, rod, 100mm, diameter 2.0mm, 99.99+%
Ni
GF45193697
Nickel, rod, 100mm, diameter 2.0mm, annealed, 99%
Ni
GF11517948
Nickel, rod, 100mm, diameter 20.0mm, 99%
Ni
GF39205931
Nickel, rod, 100mm, diameter 25.4mm, 99.98%
Ni
GF69167920
Nickel, rod, 100mm, diameter 3.0mm, 99%
Ni
GF70332870
Nickel, rod, 100mm, diameter 3.3mm, 99.99+%
Ni
GF99799968
Nickel, rod, 100mm, diameter 30mm, 99%
Ni
GF90754241
Nickel, rod, 100mm, diameter 40mm, 99%
Ni
GF20561385
Nickel, rod, 100mm, diameter 5.0mm, 99.99+%
Ni
GF90932823
Nickel, rod, 100mm, diameter 6.35mm, 99%
Ni
GF50593002
Nickel, rod, 100mm, diameter 9.5mm, 99%
Ni
GF68355085
Nickel, rod, 200mm, diameter 1.5mm, 99.99+%
Ni
GF66250256
Nickel, rod, 200mm, diameter 10.0mm, 99.99+%
Ni
GF69613825
Nickel, rod, 200mm, diameter 12.0mm, 99%
Ni
GF49774034
Nickel, rod, 200mm, diameter 12.7mm, 99.98%
Ni
GF97332554
Nickel, rod, 200mm, diameter 16mm, 99.99+%
Ni
GF58660746
Nickel, rod, 200mm, diameter 2.0mm, 99.99+%
Ni
GF71556958
Nickel, rod, 200mm, diameter 2.0mm, annealed, 99%
Ni
GF39742577
Nickel, rod, 200mm, diameter 20.0mm, 99%
Ni
GF11244259
Nickel, rod, 200mm, diameter 25.4mm, 99.98%
Ni
GF51944665
Nickel, rod, 200mm, diameter 3.0mm, 99%
Ni
GF91609555
Nickel, rod, 200mm, diameter 3.3mm, 99.99+%
Ni
GF95866824
Nickel, rod, 200mm, diameter 30mm, 99%
Ni
GF44280974
Nickel, rod, 200mm, diameter 40mm, 99%
Ni
GF70172838
Nickel, rod, 200mm, diameter 5.0mm, 99.99+%
Ni
GF17475903
Nickel, rod, 200mm, diameter 6.35mm, 99%
Ni