EMAIL THIS PAGE TO A FRIEND
Cell cycle (Georgetown, Tex.)
The CDK-activating kinase Cdk7: taking yes for an answer.
PMID 24036541
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV03009
Anti-CDK7 antibody produced in rabbit, IgG fraction of antiserum