EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Functionalization of benzylic C(sp(3))-H bonds of heteroaryl aldehydes through N-heterocyclic carbene organocatalysis.
PMID 24038663
Abstract
Aryl aldehyde activation: Oxidative activation of 2-methylindole-3-carboxaldehyde (I) through N-heterocyclic carbene (NHC) organocatalysis generates heterocyclic ortho-quinodimethane (II) as a key intermediate. This intermediate then undergoes formal [4+2] cycloaddition with trifluoromethyl ketones or isatins to form polycyclic lactones containing a quaternary carbon center.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
262439
2-Methylindole-3-carboxaldehyde, 97%
C10H9NO